ContactUsLinkComponent
Everbrite Mounting Med W/Dapi
EverBrite™ Fluorescence Antifade Mounting Media and CoverGrip™ Coverslip Sealant, Biotium
# de catalogue 89411-106
Fournisseur:  Biotium
Numéro CAS:  
Everbrite Mounting Med W/Dapi
EverBrite™ Fluorescence Antifade Mounting Media and CoverGrip™ Coverslip Sealant, Biotium
# de catalogue 89411-106
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    EverBrite™ Mounting Medium with DAPI
  • Size:
    10 mL
  • Cat. no.:
    89411-106
  • Supplier No.:
    23002
Spécifications
A propos de cet article
EverBrite™ Mounting Media are anti-fade mountants for preserving fluorescence during microscopy
EverBrite™ Mounting Medium is a wet-set medium, while EverBrite™ Hardset Mounting Medium cures overnight to form a hard, permanent seal with the coverslip, eliminating the need to seal coverslip edges with nail polish or other sealants. Both formulations are available with DAPI for blue fluorescent nuclear counterstaining without an additional staining step.
EverBrite™ Mounting Media are anti-fade mountants for preserving fluorescence during microscopy
EverBrite™ Mounting Medium is a wet-set medium, while EverBrite™ Hardset Mounting Medium cures overnight to form a hard, permanent seal with the coverslip, eliminating the need to seal coverslip edges with nail polish or other sealants. Both formulations are available with DAPI for blue fluorescent nuclear counterstaining without an additional staining step.
EverBrite™ Mounting Medium is a wet-set medium, while EverBrite™ Hardset Mounting Medium cures overnight to form a hard, permanent seal with the coverslip, eliminating the need to seal coverslip edges with nail polish or other sealants. Both formulations are available with DAPI for blue fluorescent nuclear counterstaining without an additional staining step.
CoverGrip™ Coverslip Sealant is an organic adhesive medium formulated specifically for sealing the edges of wet-mounted coverslips. Unlike nail polish, CoverGrip™ does not contain alcohol, which can diffuse into aqueous mounting medium and affect sample fluorescence. CoverGrip™ is made with limonene, a pleasant-smelling natural hydrocarbon made from citrus peels that is more environmentally friendly and less toxic than other organic solvents. CoverGrip™ is supplied in a 15 mL brush cap bottle for ease of application, or in 100 mL dropper-top refill bottles.
EverBrite™ Mounting Media prevent rapid photobleaching of fluorescein, rhodamine, and cyanine based dyes and are optimally formulated for use with Biotum’s CF™ dye conjugates. Unlike Vectashield®, EverBrite™ mounting media is compatible with Cy® dyes such as Cy®2, Cy®3, and Cy®5.
Customers Who Bought This Also Bought