Order Entry
ContactUsLinkComponent
Fish Gelatin Blocking Buffer
Buffers and Accessories, Biotium
# de catalogue 89411-096
Fournisseur:  Biotium
Numéro CAS:  
Fish Gelatin Blocking Buffer
Buffers and Accessories, Biotium
# de catalogue 89411-096
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    10X Fish Gelatin Blocking Agent
  • Size:
    100 mL
  • Cat. no.:
    89411-096
  • Supplier No.:
    22010

Spécifications

A propos de cet article

Commonly used buffers and accessories for microbiology, cell biology and microscopy.

  • Ready-to-use buffers formulated for immunofluorescence staining of intracellular antigens for analysis by flow cytometry
  • Fish gelatin blocking reagent for Western blotting and immunostaining
  • Bovine serum albumin (BSA) is commonly used as a stabilizing agent for proteins and enzymes
  • Dilute solutions of antibody
  • Blocking agent to reduce non-specific binding

Fish gelatin is commonly used in blocking solutions to minimize background noise in immunodetection procedures such as Western blotting and immunostaining. Fish gelatin can be used in blocking buffers to block non-specific binding sites on positively charged nylon or PVDF membranes or for blocking and dilution of antibodies during immunostaining procedures. Unlike BSA or milk, fish gelatin does not contain IgG or serum proteins that could cross-react with mammalian antibodies.

Ultrafiltration vials with MWCO=10K are recommended for ultrafiltration of antibodies and other proteins larger than 30 kDa. Vials with MWCO=3K can be used for ultrafiltration of proteins larger than 10 kDa. Ultrafiltration can be used to rapidly and easily remove small molecules such dyes, biotin, Tris, glycerol and glycine from antibody solutions, or to concentrate antibodies prior to conjugation.

Activated Charcoal Decontamination Bags retain ethidium bromide, GelRed™ and GelGreen™ efficiently in a convenient bag for decontamination of used gel staining solutions. Each bag has a total capacity of at least 10 mg ethidium bromide.
Mini glass tubes offer higher sensitivity than plastic tubes when used with Biotium’s AccuLite™ Mini-Fluorometers.

Customers Who Bought This Also Bought