Ec-25P 25 feet Extension Cable
# de catalogue 89409-836
Fournisseur: Monarch Instrument
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Fournisseur: VWR International
109,33 $ to 364,87 $
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
377,44 $ to 992,90 $
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
194,14 $ to 694,12 $