T-5 Reflective Tape 5 feet Roll .5 Inch w
# de catalogue 89409-830
Fournisseur: Monarch Instrument
Customers Who Bought This Also Bought
Formaldehyde 37% in aqueous solution stabilized, VWR®, Reagent Grade
Fournisseur: VWR International
$128.91 to $322.93
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Fournisseur: VWR International
$109.33 to $364.87
Sodium hypochlorite 12.5% in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$99.85 to $385.36
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$194.14 to $694.12