Rols-P Remote Optical Sensor
# de catalogue 89409-826
Fournisseur: Monarch Instrument
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
$881.80 to $2,807.56
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Fournisseur: VWR International
$458.88 to $1,237.78
Sodium sulfate, anhydrous ≥99.0%, granular, Tracepur® 10-60 mesh, Supelco®
Fournisseur: MilliporeSigma
$1,352.49
Checker Plus® pH Tester, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
$250.86
Dichloromethane ≥99.8% (by GC, corrected for water content) stabilized, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
$795.94 to $2,497.03