Tryptic Soy Broth (TSB), Hardy Diagnostics
# de catalogue 89407-988
Fournisseur: Hardy Diagnostics
Product available on GSA Advantage®
Numéro CAS:
Spécifications
- Packaging:1000 mL Polycarbonate Bottle
- Description:TSB, USP
- Size:1000 mL
- Storage conditions:–2-30°C
- Minimum Order Quantity:—
- Supplier No.:U67
Spécifications
A propos de cet article
For the general cultivation of microorganisms.
Delivery: Ships Ground Only.
Customers Who Bought This Also Bought