ContactUsLinkComponent
Serological Pipette,5Ml,Sterile,Single Paper Pack Cs200
Nunc™ Serological Pipettes, Thermo Scientific
# de catalogue 89408-566
Serological Pipette,5Ml,Sterile,Single Paper Pack Cs200
Nunc™ Serological Pipettes, Thermo Scientific
# de catalogue 89408-566
Spécifications
  • Capacity:
    5 ml
  • Sterility:
    Sterile
  • Material:
    PS
  • Cat. no.:
    89408-566
  • Supplier No.:
    170355
Spécifications
A propos de cet article
Select size and packaging options to advance your research with complete confidence when using Nunc™ Serological Pipettes
These accurate, disposable, plastic pipettes are supplied plugged and sterilized.
Customers Who Bought This Also Bought