MBP 101 to 1000 µl Pipette Tips, Thermo Scientific
# de catalogue 89404-804
Fournisseur: Thermo Scientific
Spécifications
- Volume:101 - 1000 µl
- Tip Color:Blue
- Packed:Bagged
- Cat. no.:89404-804
- Supplier No.:111-11
Spécifications
A propos de cet article
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product.
- Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals
- A trace metal certificate is available upon request
- A mold release agent is NOT used in the manufacturing of these pipette tips
Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-sterilized product is not certified as pyrogen-free).
Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation.
Ordering information: Certificate of irradiation and Certificate of analysis are available upon request.
Customers Who Bought This Also Bought