ContactUsLinkComponent
Pipette Tip 1000-5000Ul Nat Bag
Universal Fit Pipette Tips, Molecular Bioproducts
Catalog # 89404-784
Supplier:  Thermo Scientific
Universal Fit Pipette Tips, Molecular Bioproducts
Catalog # 89404-784
Supplier:  Thermo Scientific
Specifications
  • Volume:
    1000 - 5000 µL
  • Tip Color:
    Natural
  • Cat. no.:
    89404-784
  • Tip Style:
    Universal Fit, Macro Volume
  • Supplier No.:
    096