ContactUsLinkComponent
NITRATE QUANT KIT 100TST/EA
MQuant™ Nitrate Test Strips, Colorimetric, 10 - 25 - 50 - 100 - 250 - 500 mg/L NO₃⁻, MilliporeSigma
# de catalogue CA1.10020.0001
Fournisseur:  MilliporeSigma
Numéro CAS:  
MQuant™ Nitrate Test Strips, Colorimetric, 10 - 25 - 50 - 100 - 250 - 500 mg/L NO₃⁻, MilliporeSigma
# de catalogue CA1.10020.0001
Fournisseur:  MilliporeSigma
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Chemical method:
    Griess' reaction
  • Description:
    Test with Strips
  • Cat. no.:
    CA1.10020.0001
  • Range:
    10-25-50-100-250-500 mg/l NO₃
  • Supplier No.:
    1.10020.0001
  • No. of Tests:
    100 strips
Spécifications
A propos de cet article
The nitrate test can be used for monitoring of drinking water, process water and wastewater, as well as aquarium water where limits can quickly be exceeded
MilliporeSigma # 1.10020.0001 replaces and is identical to MilliporeSigma # 10020-1
It can also be used for determine nitrate in food e.g. potatoes.
BodyTextAccessories: Analyze your nitrate test strips with the reference card and with the free MQuant® StripScan app. The brilliant and colorfast reference colors allow for robust measurement results even under changing lighting conditions.The StripScan app allows the customer to analyze the MQuant® test strips with your smartphone for fast, convenient, and precise results which can be easily graphically presented and shared.
Customers Who Bought This Also Bought