Spécifications
- Chemical method:Griess' reaction
- Description:Test with Strips
- Cat. no.:CA1.10020.0001
- Range:10-25-50-100-250-500 mg/l NO₃
- Supplier No.:1.10020.0001
- No. of Tests:100 strips
Spécifications
A propos de cet article
The nitrate test can be used for monitoring of drinking water, process water and wastewater, as well as aquarium water where limits can quickly be exceeded
MilliporeSigma # 1.10020.0001 replaces and is identical to MilliporeSigma # 10020-1
It can also be used for determine nitrate in food e.g. potatoes.