ContactUsLinkComponent
Sulfite Test Method: Colorimetric With Test Strips 10 - 40 - 80 - 180 - 400 mg/L SO32- MQuant™, MilliporeSigma
Catalogue # CA1.10013.0001
Fournisseur:  MilliporeSigma
Numéro CAS:  
Sulfite Test Method: Colorimetric With Test Strips 10 - 40 - 80 - 180 - 400 mg/L SO32- MQuant™, MilliporeSigma
Catalogue # CA1.10013.0001
Fournisseur:  MilliporeSigma
Supplier Number:  1.10013.0001
Numéro CAS:  
Spécifications
  • Chemical method:
    Nitroprusside / Zn-hexacyanoferrate
  • Description:
    Test with Strips
  • Cat. no.:
    CA1.10013.0001
  • Range:
    10-40-80-180-400 mg/l SO₃
  • Supplier No.:
    1.10013.0001
  • No. of Tests:
    100 Strips
Spécifications
À propos de cet article
The sulfite test is suitable for food analysis, particularly in the testing of wine and fruit juices, sulfurated foods (e.g
MilliporeSigma # 1.10013.0001 replaces and is identical to MilliporeSigma # 10013-1
fresh and dried fruits), fish shellfish, crustaceans and minced meat products.
Frequently Bought Together