Sulfite Test Method: Colorimetric With Test Strips 10 - 40 - 80 - 180 - 400 mg/L SO32- MQuant™, MilliporeSigma
Catalogue # CA1.10013.0001
Fournisseur: MilliporeSigma
Numéro CAS:
Spécifications
- Chemical method:Nitroprusside / Zn-hexacyanoferrate
- Description:Test with Strips
- Cat. no.:CA1.10013.0001
- Range:10-40-80-180-400 mg/l SO₃
- Supplier No.:1.10013.0001
- No. of Tests:100 Strips
Spécifications
À propos de cet article
The sulfite test is suitable for food analysis, particularly in the testing of wine and fruit juices, sulfurated foods (e.g
MilliporeSigma # 1.10013.0001 replaces and is identical to MilliporeSigma # 10013-1
fresh and dried fruits), fish shellfish, crustaceans and minced meat products.
Frequently Bought Together