ContactUsLinkComponent
NITRITE QUANT KIT 100TST/EA
MQuant™ Nitrite Test Strips, MilliporeSigma
# de catalogue CA1.10007.0001
Fournisseur:  MilliporeSigma
Numéro CAS:  
MQuant™ Nitrite Test Strips, MilliporeSigma
# de catalogue CA1.10007.0001
Fournisseur:  MilliporeSigma
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Chemical method:
    Griess' reaction
  • Description:
    Test with Strips
  • Cat. no.:
    CA1.10007.0001
  • Range:
    2-5-10-20-40-80 mg/L NO₂
  • Supplier No.:
    1.10007.0001
  • No. of Tests:
    100 strips
Spécifications
A propos de cet article
MQuant™ (MQ) test strips can be considered as a mobile laboratory on a few square millimetres of plastic strip
  • Easy to handle: just dip, wait and measure visually against colour scale
  • Safe: no auxiliary reagents necessary
  • Immediate results within 2 minutes enables fast decisions
  • Portable: can be used inside or outside the laboratory
MilliporeSigma # 1.10022.0001 replaces and is identical to MilliporeSigma # 10022-1. MilliporeSigma # 1.10007.0001 replaces and is identical to MilliporeSigma # 10007-1
As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, possibly leading to substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content in the reaction zones offers problem-free waste disposal.
1.0022.0001 can be used for measuring the nitrite concentration of boiler water and cooling water.
Customers Who Bought This Also Bought