Spécifications
- Chemical method:Griess' reaction
- Description:Test with Strips
- Cat. no.:CA1.10007.0001
- Range:2-5-10-20-40-80 mg/L NO₂
- Supplier No.:1.10007.0001
- No. of Tests:100 strips
Spécifications
A propos de cet article
MQuant™ (MQ) test strips can be considered as a mobile laboratory on a few square millimetres of plastic strip
- Easy to handle: just dip, wait and measure visually against colour scale
- Safe: no auxiliary reagents necessary
- Immediate results within 2 minutes enables fast decisions
- Portable: can be used inside or outside the laboratory
MilliporeSigma # 1.10022.0001 replaces and is identical to MilliporeSigma # 10022-1. MilliporeSigma # 1.10007.0001 replaces and is identical to MilliporeSigma # 10007-1
As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, possibly leading to substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content in the reaction zones offers problem-free waste disposal.
1.0022.0001 can be used for measuring the nitrite concentration of boiler water and cooling water.