Spécifications
- Chemical method:Rhodanide
- Description:Test with Strips
- Cat. no.:CA1.10002.0001
- Range:10-30-100-300-1000 mg/l Co
- Supplier No.:1.10002.0001
- No. of Tests:100 strips
Spécifications
A propos de cet article
The cobalt test can be used in the testing of waste water, electroplating baths, in the electronics industry, metal industry, in the assaying and refining of ores.
MilliporeSigma # 1.10002.0001 replaces and is identical to MilliporeSigma # 10002-1