ContactUsLinkComponent
COLORPHAST PH6.5-10 6X100/EA
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Catalog # CA1.09543.0007
Supplier:  MilliporeSigma
undefined
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Catalog # CA1.09543.0007
Supplier:  MilliporeSigma
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    pH-Indicator strips, special indicator, Non-bleeding, narrow range
  • Cat. no.:
    CA1.09543.0007
  • pH Range:
    6.5 - 10.1
  • pH Graduation:
    6.5-6.8-7.1-7.4-7.7-7.9-8.1-8.3-
    8.5-8.7-9.0-9.5-10.0
  • Resolution:
    0.3
  • Supplier No.:
    1.09543.0007
  • No. of Strips:
    600
Specifications
About this item
These strips provide a convenient method for accurate colorimetric pH indication.
The strips feature covalently bound indicator dyes so that they will not bleed even in strong alkaline solutions. Strips can be immersed in samples for extended periods, so that even weakly buffered solutions can be accurately tested without contaminating the sample. The universal strip tests the full 0–14 pH range with a sensitivity of 1 pH unit. Intermediate range indicators provide a sensitivity of 0.3–0.5 pH units and narrow range strips provide a sensitivity of 0.2–0.3 pH units.
MilliporeSigma # 1.09531.0001 replaces and is identical to MilliporeSigma # 9586-1.
MilliporeSigma # 1.09531.0007 replaces and is identical to MilliporeSigma # 9586-3.
MilliporeSigma # 1.09532.0001 replaces and is identical to MilliporeSigma # 9587-1.
MilliporeSigma # 1.09533.0001 replaces and is identical to MilliporeSigma # 9588-1.
MilliporeSigma # 1.09533.0007 replaces and is identical to MilliporeSigma # 9588-3.
MilliporeSigma # 1.09535.0001 replaces and is identical to MilliporeSigma # 9590-1.
MilliporeSigma # 1.09535.0007 replaces and is identical to MilliporeSigma # 9590-3.
MilliporeSigma # 1.09540.0001 replaces and is identical to MilliporeSigma # 9580-1.
MilliporeSigma # 1.09541.0001 replaces and is identical to MilliporeSigma # 9581-1.
MilliporeSigma # 1.09542.0001 replaces and is identical to MilliporeSigma # 9582-1.
MilliporeSigma # 1.09542.0007 replaces and is identical to MilliporeSigma # 9582-3.
MilliporeSigma # 1.09543.0001 replaces and is identical to MilliporeSigma # 9583-1.
MilliporeSigma # 1.09543.0007 replaces and is identical to MilliporeSigma # 9583-3.
MilliporeSigma # 1.09545.0001 replaces and is identical to MilliporeSigma # 9585-1.
MilliporeSigma # 1.09584.0001 replaces and is identical to MilliporeSigma # 9578-1.
BodyTextAccessories: Analyze your nitrate test strips with the reference card and with the free MQuant® StripScan app. The brilliant and colorfast reference colors allow for robust measurement results even under changing lighting conditions.The StripScan app allows the customer to analyze the MQuant® test strips with your smartphone for fast, convenient, and precise results which can be easily graphically presented and shared.
Frequently Bought Together