ContactUsLinkComponent
Phosphorus, Reactive, Test N Tube* Reagent Set
Phosphorus (Reactive) TNT Reagent Set, Low Range, Hach
# de catalogue CA97026-534
Fournisseur:  Hach
Numéro CAS:  
Phosphorus, Reactive, Test N Tube* Reagent Set
Phosphorus (Reactive) TNT Reagent Set, Low Range, Hach
# de catalogue CA97026-534
Fournisseur:  Hach
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Chemical method:
    Ascorbic acid
  • Description:
    Phosphorus (Reactive) TNT Reagent Set, Low Range
  • Method:
    8048 - Ascorbic Acid
  • Instrument:
    All except DR/820, PC II
  • Cat. no.:
    CA97026-534
  • Range:
    0.06 - 5.00 mg/l PO₄
  • Supplier No.:
    2742545
  • No. of Tests:
    25 - 50
Spécifications
A propos de cet article
For determination of Reactive Phosphorus (ortho-phosphate) by the PhosVer® 3 Ascorbic Acid Test 'N Tube™ method.
-Test 'N Tube products provide safe, convenient testing
-Easy ordering
  • Test 'N Tube products provide safe, convenient testing
  • Easy ordering
Hach Method 8048. Range: 0.06 - 5.00 mg/L PO4. Set includes: PhosVer® 3 Phosphate Reagent Powder Pillows and Reactive Phosphorus Test 'N Tube Dilution Vials for up to 50 tests.
Customers Who Bought This Also Bought