Spécifications
- Chemical method:Ascorbic acid
- Description:Phosphorus (Reactive) TNT Reagent Set, Low Range
- Method:8048 - Ascorbic Acid
- Instrument:All except DR/820, PC II
- Cat. no.:CA97026-534
- Range:0.06 - 5.00 mg/l PO₄
- Supplier No.:2742545
- No. of Tests:25 - 50
Spécifications
A propos de cet article
For determination of Reactive Phosphorus (ortho-phosphate) by the PhosVer® 3 Ascorbic Acid Test 'N Tube™ method.
-Test 'N Tube products provide safe, convenient testing
-Easy ordering
- Test 'N Tube products provide safe, convenient testing
- Easy ordering
Hach Method 8048. Range: 0.06 - 5.00 mg/L PO4. Set includes: PhosVer® 3 Phosphate Reagent Powder Pillows and Reactive Phosphorus Test 'N Tube Dilution Vials for up to 50 tests.