ContactUsLinkComponent
Tester Bart Srb Pk/9
BART Test for Sulfate-Reducing Bacteria, Hach
# de catalogue CA97026-322
Fournisseur:  Hach
Numéro CAS:  
BART Test for Sulfate-Reducing Bacteria, Hach
# de catalogue CA97026-322
Fournisseur:  Hach
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Cat. no.:
    CA97026-322
  • Supplier No.:
    2432409
Spécifications
A propos de cet article
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria. Sulfate Reducing Bacteria (SRB) are one of the most important agents in biofouling and can cause corrosion, clogging, fouling of the water, and increased hygiene risks.
Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria.
Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria.
Easy to use, requiring no elaborate or costly equipment and no specialized training. Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment.
Customers Who Bought This Also Bought