ContactUsLinkComponent
Tough Spots label In Coff 1000/Rl
Adhesive Labels on Rolls, Round, Electron Microscopy Sciences
# de catalogue 102681-366
Tough Spots label In Coff 1000/Rl
Adhesive Labels on Rolls, Round, Electron Microscopy Sciences
# de catalogue 102681-366
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    Coffee
  • Description:
    Labels on rolls
  • Cat. no.:
    102681-366
  • Packaging:
    1000 labels/roll
  • Dimensions:
    0.375"
  • Supplier No.:
    77580-CO
Spécifications
A propos de cet article
Tough-Spots® pre-cut peel-off round labels are used on microcentrifuge tube tops. The PVC labels are easy to write on and accept writing from most lab markers including solvent resistant pens and teeny writers.
  • In sheets (for laser printer) or on a roll
  • Select from 11 colors
  • A variety of sizes, colors and formats ensures the right label for the job
Ordering information: Rainbow includes blue, green, orange, red, and yellow.
Delivery: 5 Across Tough-Spots® come in a dispensing box that assists in labeling individual tubes used in small experiments. White or assorted roll with 1000 spots each of blue, green, lavender, red and yellow.
Customers Who Bought This Also Bought