Flash Column Spherical Amide(HILIC) 20-45 um 100 4g 20/Pack
# de catalogue 89401-950
Fournisseur: Agela Technologies
Flash Column Spherical Amide(HILIC) 20-45 um 100 4g 20/Pack
# de catalogue 89401-950
Fournisseur: Agela Technologies
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Fournisseur: VWR International
$109.33 to $364.87
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Fournisseur: MilliporeSigma
$1,313.27 to $5,149.78
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Fournisseur: Air-Tite
$77.41 to $1,124.20