Order Entry
ContactUsLinkComponent
Pouring Ring Gl45Pp Clear Pk10
Pouring Rings, Chemglass
Catalog # 30623-040
Supplier:  Chemglass
Small Business Enterprise
Pouring Ring Gl45Pp Clear Pk10
Pouring Rings, Chemglass
Catalog # 30623-040
Supplier:  Chemglass
Small Business Enterprise

Specifications

  • Description:
    Pouring Ring, GL-45, Clear, Polypropylene
  • Cat. no.:
    30623-040
  • Supplier No.:
    CLS-1482-17

Specifications

About this item

These pour rings are made of high temperature ETFE (Ethylene Tetrafluoroethylene).

  • Steam autoclavable to 140 °C
  • Pouring rings are polypropylene
  • Max temperature is 180 °C
Customers Who Bought This Also Bought