Solid Caps, GL-Threads, Polypropylene, Chemglass
# de catalogue 30623-020
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Color:Orange
- Description:Solid Caps
- Cat. no.:30623-020
- Thread Size:GL45
- Supplier No.:CLS-1480-16
Spécifications
A propos de cet article
These solid polypropylene caps can be autoclaved in steam to 140 °C.
Customers Who Bought This Also Bought