ContactUsLinkComponent
3Port Harvest Tube For 3L Vsa
Accessories for Cylindrical Vessels with Vertical Side Arm, Chemglass
# de catalogue 30622-900
Fournisseur:  Chemglass
Small Business Enterprise
3Port Harvest Tube For 3L Vsa
Accessories for Cylindrical Vessels with Vertical Side Arm, Chemglass
# de catalogue 30622-900
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Description:
    Harvest tube, 3-port with two ¼" and one 0.09375" tubes
  • Cat. no.:
    30622-900
  • For Use With:
    3000 ml bioreactor vessel
  • Supplier No.:
    CLS-1437-70