ContactUsLinkComponent
Chemrxnhub 5L Process Sys Comp
ChemRxnHub™ Reactor System, Jacketed, Chemglass
# de catalogue 30622-588
Fournisseur:  Chemglass
Small Business Enterprise
Chemrxnhub 5L Process Sys Comp
ChemRxnHub™ Reactor System, Jacketed, Chemglass
# de catalogue 30622-588
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • W×D×H:
    15.25×17.25×40" (Stan)
  • Description:
    ChemRxnHub Process System, Complete with Glassware, Stand, OHS 100 Advance Motor and Manifold/Drain System
  • Cat. no.:
    30622-588
  • Supplier No.:
    CG-1949-X-5LM
Spécifications
A propos de cet article
ChemRxnHub™ process systems are complete with benchtop stand, overhead stirrer and glassware.
  • Available with the optional manifold connection and heat transfer fluid draining system
Ordering information: For a list of the standard components supplied with complete Chemrxnhub systems or for custom glassware setups; contact your Avantor representative.
Customers Who Bought This Also Bought