ContactUsLinkComponent
Chemrxnhub 300Ml Processsystem
ChemRxnHub™ Systems, Jacketed, 300 ml, Jacketed Lab Reactor, Chemglass
# de catalogue 30622-578
Fournisseur:  Chemglass
Small Business Enterprise
Chemrxnhub 300Ml Processsystem
ChemRxnHub™ Systems, Jacketed, 300 ml, Jacketed Lab Reactor, Chemglass
# de catalogue 30622-578
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Description:
    ChemRxnHub Process System Complete with Glassware, Stand, and OHS 100 Advance Motor
  • Cat. no.:
    30622-578
  • Supplier No.:
    CG-1949-X-300
Spécifications
A propos de cet article
ChemRxnHub™ process systems are complete with benchtop stand, overhead stirrer and glassware.
Available with the optional manifold connection and heat transfer fluid draining system. The unique design of the ChemRxnHub™ support stand and clamping system allows for quick and easy installation/removal of glassware and fits into most benchtop hoods.
Customers Who Bought This Also Bought