Hydrochloric acid 36.5 - 38.0% ACS
# de catalogue CA11020-884
Fournisseur: Anachemia
Numéro CAS: 7647-01-0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Poly bottle
- Size:2.5 L
- Cat. no.:CA11020-884
- Supplier No.:46414-463
Spécifications
Résultats des tests de spécification
- Assay (as HCl):36.5 - 38.0%
- Appearance:Passes Test
- Colour (APHA):10 max.
- Residue after ignition:5 ppm max.
- Bromide (Br):0.005% max.
- Sulfate (SO4):1 ppm max.
- Sulfite (SO3):1 ppm max.
- Extractable organic substances:Passes Test (about 5 ppm)
- Free Chlorine (Cl):1 ppm max.
- Ammonium (NH4):3 ppm max.
- Arsenic (As):0.01 ppm max.
- Heavy metals (as Pb):1 ppm max.
- Iron (Fe):0.2 ppm max.
- Hydrochloric acid 36.5 - 38.0% ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Hydrochloric acid solution
Point d'ébullition:110 °C (1013 hPa)
Point de Fusion:–30 °C
Densité:1.18 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 110 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –30 °C
:Flash Pt:
:Flash Pt:
:M.W. 36.46 g/mol
:36.46
Température d'entreposage:Ambient
EINECS:231-595-7
ONU:1789
:8
:II
:GHS05
:GHS07
Customers Who Bought This Also Bought