Calcium Standards, CRM for AA and ICP, Assurance Grade, SPEX CertiPrep®
# de catalogue CA11020-444
Fournisseur: Spex Certiprep
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Concentration:1000 µg/ml
- Element:Calcium
- Matrix:2% HNO₃
- Packaging:Plastic Bottle
- Size:125 ml
- Supplier No.:PLCA2-2Y
Spécifications
A propos de cet article
- Traceable to NIST
- Uncertainty of standards is reported on the Certificate of Analysis
- Manufactured by experienced chemists to ensure highest quality standards
Caution: 1000 µg/ml = 1000 ppm.
Customers Who Bought This Also Bought