Order Entry
ContactUsLinkComponent
BAG general use PP 6X10IN CS100
Polypropylene Bags, Dynalon
Catalog # 30620-112
Supplier:  Dynalon
BAG general use PP 6X10IN CS100
Polypropylene Bags, Dynalon
Catalog # 30620-112
Supplier:  Dynalon

Specifications

  • Thickness:
    1.5 mil
  • Cat. no.:
    30620-112
  • W×L:
    152×254 mm (6×10")
  • Supplier No.:
    628525

Specifications

About this item

Transparent general-use plastic bags.

  • 0.0015" thick
  • Polypropylene construction

Economical general-use plastic bags are especially useful as specimen, sample, and freezing bags and are ideal for storing and shipping items.

Customers Who Bought This Also Bought