SP Bel-Art Clavies™ Autoclavable Bags, Bel-Art Products, a part of SP
Catalog # 30617-608
Supplier: Bel-Art Products, a Part of SP
Specifications
- Dimensions:30×61 cm (12×24")
- Cat. no.:30617-608
- Supplier No.:131851224
Specifications
About this item
Clavies® bags offer maximum protection and performance for autoclavibility.
- Steam autoclavable at 135 °C (275 °F)
The durable 0.05 mm thick polypropylene bag is filled with used labware such as Petri dishes, membrane filters, culture plates, pipettes, and more, then autoclaved and discarded.
Customers Who Bought This Also Bought