ContactUsLinkComponent
Vwr Buf Blood Bank Saline 2.5G
VWR® Blood Bank Saline Solutions
# de catalogue 89370-100
Fournisseur:  VWR International
Numéro CAS:  
Vwr Buf Blood Bank Saline 2.5G
VWR® Blood Bank Saline Solutions
# de catalogue 89370-100
Fournisseur:  VWR International
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    VWR® Buffered Blood Bank Saline
  • Size:
    2.5 gal.
Spécifications
A propos de cet article
Blood bank saline is pH-controlled for a pH between 6.5 and 7.0.
  • pH-controlled
  • Enhances weak agglutination reactions
  • Stable without preservatives
Buffered saline is tightly controlled to a pH range of 7.0 to 7.2 and is specially designed to enhance weak agglutination reactions in blood bank procedures. Both saline and buffered saline remain stable without preservatives.
Customers Who Bought This Also Bought