Specifications
- Description:VWR® Blood Bank Saline
- Size:2.5 gal.
- Cat. no.:89370-096
Specifications
About this item
Blood bank saline is pH-controlled for a pH between 6.5 and 7.0.
- pH-controlled
- Enhances weak agglutination reactions
- Stable without preservatives
Buffered saline is tightly controlled to a pH range of 7.0 to 7.2 and is specially designed to enhance weak agglutination reactions in blood bank procedures. Both saline and buffered saline remain stable without preservatives.