Alcohol 100%, VWR®, Reagent Grade for histology
# de catalogue 89370-084
Fournisseur: VWR International
Product available on GSA Advantage®
Numéro CAS: 64-17-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Size:1 gal.
Spécifications
Résultats des tests de spécification
- Components:
- Ethanol:89.5-91.5%
- Methanol:4.0-5.0%
- Isopropanol:4.5-5.5%
- Characteristics:
- Methanol and Ethyl Alcohol (SDA 3A 200 Proof) (v/v):94.0 – 96.0%
- Isopropyl Alcohol (2-Propanol) (v/v):4.0 – 6.0%
- Water:max 0.2%
- Residue After Evaporation:max 10 ppm
- Appearance:Clear
- Specific Gravity:0.7902-0.7912 @ 20° C
- Color (Pt-Co):10 max
- Odor:Pass
- Titrable Acid:0.0003meq/g
- Titrable Base:0.0002meq/g
- Flourescent Background:Pass
- Identification:Pass
- Substances Reducing KMnO4:Pass
- Solubility in Water:Pass
- Refractive Index @ 25°C:1.3580-1.3610
- Alcohol 100%, VWR®, Reagent Grade for histology:
Résultats des tests de spécification
A propos de cet article
Synonymes: Reagent alcohol
:Alcool
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:100%
:100
:Point d'ébullition:
:Boiling Pt:
:Point de fusion:
:Point eclaire:
:Flash Pt:
ONU:1987
:3
:II
:GHS02
:GHS08
:GHS07
Température d'entreposage:Ambient
Customers Who Bought This Also Bought