Hydrion® Quat Test Papers, Micro Essential Laboratory
# de catalogue 89370-914
Fournisseur: Micro Essential
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Quat Test Papers Dispenser, 0–1000 ppm
- Cat. no.:89370-914
- Measurement Range:0 - 200 - 400 - 600 - 800 - 1000ppm
- Supplier No.:QC-1001
Spécifications
A propos de cet article
A quick, reliable, easy-to-use, way to test quaternary ammonium based sanitizers in food service establishments, as required by federal, state, and local food safety codes.
Delivery: Kit consists of a 15' x 5/32" roll of premium test paper in a handy Hydrion dispenser with a matching color chart.
Customers Who Bought This Also Bought