Dispenser Refill Packs for Sentino® Filter Dispenser, Cytiva (Formerly Pall Lab)
# de catalogue CA30617-202
Fournisseur: Cytiva
Spécifications
- Pore size:0.45 µm
- Description:Dispenser Refill Pack, Metricel® Black Membrane
- Material:Modified Polyethersulfone
- Cat. no.:CA30617-202
- Supplier No.:68124
Spécifications
A propos de cet article
These membranes have been designed specifically for use with the Sentino® automatic filter dispenser.
- Ideal for high volume laboratories
- Eliminate the time and waste of individually sealed packs of membrane
Delivery: Each box contains two additional discs for loading the dispenser.
Customers Who Bought This Also Bought