ContactUsLinkComponent
CELL COUNTER BLOOD 5-KEY
Blood Cell Counter, Bal Supply
# de catalogue 89370-064
Fournisseur:  Tip Industries
CELL COUNTER BLOOD 5-KEY
Blood Cell Counter, Bal Supply
# de catalogue 89370-064
Fournisseur:  Tip Industries
Spécifications
  • Description:
    Counter with Five Counting Units
  • Cat. no.:
    89370-064
  • Supplier No.:
    505C
Spécifications
A propos de cet article
Counter with two counting units has single reset knob where as counter with five and eight counting units have reset knobs on each side.
Counting units have a range of 0 to 999. Removable tabs provide an area for identification of the classification being counted.
Customers Who Bought This Also Bought