Polysorbate 80 solution, VWR Chemicals BDH®
# de catalogue BDH7781-2
Fournisseur: VWR International
Numéro CAS: 9005-65-6
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber
- Size:500 ml
- Supplier No.:BDHVBDH7781-2
Spécifications
Résultats des tests de spécification
- Appearance/color:Clear, yellow to dark yellow liquid
- Identification:Passes test
- Acid value:2.0 max.
- Hydroxyl value:65 - 80
- Saponification value:45 - 55
- Specific gravity (25°C/25 °C):1.060 - 1.090
- Viscosity (25 °C):300 - 500 cSt
- Heavy metals (as Pb):10 ppm max.
- Residue after ignition:0.25 % max.
- Water:3.0% max.
- Residual solvents:Passes test
- Polysorbate 80 solution, VWR Chemicals BDH®:
Résultats des tests de spécification
A propos de cet article
Synonymes: Sorbitan monooleate, ethoxylated,Polyoxyethylene-80-Sorbitan Monolaurate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 100 °C
:Flash Pt:
:Flash Pt: 113 °C (closed cup)
:100
:113
Customers Who Bought This Also Bought