ContactUsLinkComponent
VISTARAK* Sterile Low Retention Tips
Disposable Tips for MLA and Ovation Pipettors, VistaLab
# de catalogue 89368-682
Fournisseur:  VistaLab
VISTARAK* Sterile Low Retention Tips
Disposable Tips for MLA and Ovation Pipettors, VistaLab
# de catalogue 89368-682
Fournisseur:  VistaLab
Spécifications
  • Volume:
    250 µl
  • Packed:
    VistaRak™
  • Sterile:
    +
  • Tip Style:
    Low Retention, Small
  • Supplier No.:
    4070-2132LR
Spécifications
A propos de cet article
Disposable polypropylene tips for use with MLA or ovation pipettors.
  • Non-pyrogenic
  • Non-sterile or sterile, RNase/DNase-free
  • Filtered or non-filtered
  • Low retention available
  • Trace metal certified available
VistaLab tips are available in a full range of volumes from 2 μl to 10 ml for Macro Volume pipettors.
These disposable tips are an economical choice and are available in a wide range of sizes.
Customers Who Bought This Also Bought