Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Catalog # BDH9284-2.5KG
Supplier: VWR International
CAS Number: 497-19-8
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Poly bottle
- Size:2.5 kg
- Cat. no.:BDH9284-2.5KG
- Supplier No.:0585-VBD-2.5KG
Specifications
Specification Test Results
- Calcium:= 0.03 %
- Chloride:= 0.001 %
- Heavy Metals (as Pb):= 0.0005 %
- Insolubles:= 0.01 %
- Iron:= 0.0005 %
- Loss on Heating:= 1.0 %
- Magnesium:= 0.005 %
- Phosphate:= 0.001 %
- Potassium:= 0.005 %
- Purity:= 99.5 %
- Silica:= 0.005 %
- Sulfur Compounds:= 0.003 %
- Meets reagent specifications for testing USP/NF monographs:
- VWR Chemicals BDH®, Sodium carbonate, anhydrous ≥99.5% ACS:
Specification Test Results
About this item
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:854 °C
Density:2,53 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 1600 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: 854 °C
:Pt Eclair:|Point eclaire:
:M.W. 105,99 g/mol
Merck Index:14,08596
Storage Temperature:Température ambiante
Storage Temperature:Ambient
EINECS:207-838-8
:854
:1600
REACH:01-2119485498-19
:GHS07
:12,08739
Boiling Pt:1600 °C (1013 hPa)
Density:2.53 g/cm³ (20 °C)
:99,5
:Boiling Pt:
:Boiling Pt: 1600 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 854 °C
:Flash Pt:
:Flash Pt:
:M.W. 105.99 g/mol
Customers Who Bought This Also Bought