Sodium acetate trihydrate 99.0-101.0% ACS, VWR Chemicals BDH®
# de catalogue BDH9278-2.5KG
Fournisseur: VWR International
Numéro CAS: 6131-90-4
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Poly bottle
- Size:2.5 kg
- Supplier No.:0530-VBD-2.5KG
Spécifications
Résultats des tests de spécification
- Calcium:= 0.005 %
- Chloride:= 0.001 %
- Heavy Metals (as Pb):= 0.0005 %
- Insolubles:= 0.005 %
- Iron:= 0.0005 %
- Magnesium:= 0.002 %
- Permanganate Reducers Test:PASS
- pH (5%, Water) @25C:7.5 - 9.2
- Phosphate:= 0.0005 %
- Potassium:= 0.005 %
- Purity:99.0 - 101.0 %
- Sulfate:= 0.002 %
- Meets reagent specifications for testing USP/NF monographs.:
- VWR Chemicals BDH®, Sodium acetate trihydrate 99.0-101.0% ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Acetic acid sodium salt trihydrate
Point d'ébullition:120 - 123 °C (1013 hPa)
Point de Fusion:58 °C
Densité:1,4 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 120 à 123 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: 58 °C
:Pt Eclair:|Point eclaire:
:M.W. 136,08 g/mol
Index Merck:13,08642
Densité:1.4 g/cm³ (20 °C)
Température d'entreposage:Température ambiante
EINECS:204-823-8
:58
:12,08711
Point d'ébullition:120…123 °C (1013 hPa)
:99,0-101,0
:Boiling Pt:
:Boiling Pt: 120 to 123 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 58 °C
:Flash Pt:
:Flash Pt:
:M.W. 136.08 g/mol
:120…123 °C (1013 hPa)
Température d'entreposage:Ambient
Customers Who Bought This Also Bought