ClipTip® Pipette Tips, 50 µl
Le produit 78576-748 est un remplacement de 89348-022 et a le même emballage et les mêmes spécifications, mais peut être d'une quantité différente.
Product Details & Documents
ClipTip® pipette tips provide security with a unique and innovative interlocking technology that ensures a complete seal on every channel with minimal tip attachment and ejection force.
All ClipTip® pipette tips feature a hydrophobic inner surface which effectively prevent liquid retention. ClipTip® tips are certified free of RNase, DNase, DNA, ATP, and pyrogen contamination. The tips are available in five volumes and in multiple packaging options, including environmentally friendly reload stacks.
ClipTip® 50 pipette tips suit pipetting of 5 to 50 µl volumes.
The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip® pipette until - and only until - it is released.
Certifications: Manufactured according to ISO 9001 and ISO 13485. The F1-ClipTip® pipetting system is CE-marked in accordance with IVD Directive in Europe.
Spécifications
- Volume:5 - 50 µl
- Longueur:5.1 cm
- Tip Color:Violet
- Packed:10×96/Rack
- Filter:+
- Sterile:+
- Tip Style:ClipTip® 50, filter, racked
- For Use With:F1 - ClipTip® 5 - 50 μl, fixed 25 μl and 50 μl, 8 - channel 5 - 50 μl, and 12 - channel 5 - 50 μl
- Color Code:Purple



