Carbon Conductive Tape, Double Coated, Electron Microscopy Sciences | Avantor
Carbon Conductive Tape, Double Coated, Electron Microscopy Sciences
Double Sides Carbon Conductive Tape, 25 mm (W )x 5m (L)
Catalog # 102098-800
Fournisseur #:  77819-25
Carbon Conductive Tape, Double Coated, Electron Microscopy Sciences
Catalog # 102098-800
Carbon Conductive Tape, Double Coated, Electron Microscopy Sciences
Carbon Conductive Tape, Double Coated, Electron Microscopy Sciences
Double Sides Carbon Conductive Tape, 25 mm (W )x 5m (L)
Price
$301.85
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
EMS introduces this unique double sided carbon tape which is 260 microns in thickness
The base which is polycarbonate and 200 microns thick has on both of its sides 30 microns of conductive glue giving it a total thickness of 260 microns. The tape has one transparent liner on one side and a white thicker one on the reverse which protects the tape in shipping. These are considered to be small impurities but they may be present.
Spécifications
  • Désignation:
    Double Sides Carbon Conductive Tape, 25 mm (W )x 5m (L)