Spécifications
- Description:Premiere™ Blade #10
- Cat. no.:102097-828
- Supplier No.:72049-10
Spécifications
A propos de cet article
Premiere® Blades are made from carbon steel, individually foil wrapped and gamma radiation sterilized.
They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.