Cryogenic Grid Box, Electron Microscopy Sciences
Catalog # 102097-442
Supplier: Electron Microscopy Sciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Color:Blue
- Description:Grid Box Round with Lid
- Cat. no.:102097-442
- Supplier No.:71166-10
Specifications
About this item
These specimen grid boxes are for storing or transferring cryogenic TEM specimen grids.
- Four diamond shaped slots
- Non-static clear cover shield held in place with stainless steel screw, which is tapped in the center of the box
- Box only fits 71166-20 (Grid box square with lid)
- Available with lid, pin-type lid, or without lid
Customers Who Bought This Also Bought