ContactUsLinkComponent
Petri Dish Sqr Grid 500/Cs
Square Petri Dish, Electron Microscopy Sciences
# de catalogue 102097-178
Petri Dish Sqr Grid 500/Cs
Square Petri Dish, Electron Microscopy Sciences
# de catalogue 102097-178
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Square Petri Dish with Grid
  • Cat. no.:
    102097-178
  • Supplier No.:
    70691-CS
Spécifications
A propos de cet article
The square Petri dish with grid is consists of 36 of 13 x 13 mm squares, each 13 mm grid is marked numerically in one direction and alphabetically in the other
Made of polystyrene. Radiation sterile. This dish commonly referred to as a 100 x 15 mm dish. Free from optical distortion and sterile, these dishes are ideal for phase typing, susceptibility testing, plate counts, and probe assays.
The square Petri dish with grid is consists of 36 of 13 x 13 mm squares, each 13 mm grid is marked numerically in one direction and alphabetically in the other. This dish has four venting ribs into the underside of the lid to prevent condensation build-up.
This dish has four venting ribs into the underside of the lid to prevent condensation build-up.
Customers Who Bought This Also Bought