To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:AluminaSeal Sealing Film, for use with Cold Storage
- L×W:6×3.25"
- Material:Aluminum
- Thickness:68.6 µm
- Cat. no.:102097-052
- Supplier No.:70534-10
Spécifications
A propos de cet article
Use sealing films to eliminate well-to-well contamination, microbial and viral contamination, and sample spillage. Non breathable membranes prevent evaporation.
- Sterilized by gamma irradiation and free of cytotoxins
- Two scored end-tabsmake film positioning and removal hassle-free
- Thermal Stability to withstand a wide temperature range
- Pierceable sealing films facilitate easy removal of samples with a pipet tip
Breathe-Easy® is an adhesive, gas-permeable, cell culture membrane. It is made of 25.4 µm thick polyurethane. It is UV transparent to 300 nm and can be used in temperature range of -80 to 100 °C.
Breathe-EASIER™ is a rapid gas transmissable sealing membrane for cell growth. Breathe-Easier membranes permit rapid, consistent gas exchange of Oxygen, CO2 and water vapor for uniform cell. It is made of 139.7 µm thick non-woven rayon. It is not UV transparent and can be used in temperature range of -20 to 80 °C.
AluminaSeal™ is non-cytotoxic and sterile and is resistant to DMSO, chemicals, solvents and other common reagents. It can be used for Cold and PCR Storage in a temperature range of -80 to 110 °C. It is made of 68.6 µm aluminum and is not gas permeable or UV transparent.
HotSeal™ is a PCR sealing film for use in the temperature range of -40 to 125 °C. It is made of 58.4 µm polyester and is not gas permeable or UV transparent.