Specifications
- Color:Clear
- Description:Petriseal
- Size:0.50"x108'
- Cat. no.:102097-032
- Supplier No.:70500-C
Specifications
About this item
Ideal sealing tapes for petri dishes, microplates, lids, caps, and other lab items.
- Enjoy all the benefits of Petriseal and Containerseal in a new wider format
- Containerseal XL makes it easier to seal large containers and bottles and to cover wider gaps
Stretches, conforms, and seals the irregular perimeter of microplates or petri dishes. Waterproof and chemical resistant. Tapes and adhesive resist most solvents and caustic agents. Temperature resistant, seal remains flexible in moderate freezer temperatures and withstands intermittent exposure to 100°C.