To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Sterility:Sterile
- Description:25 mL Reservoir
- Packed:Individually Wrapped
- Cat. no.:102096-976
- Supplier No.:70496-30
Specifications
About this item
Reagent reservoirs are disposable and available in both sterile and non sterile varieties.
- Unique "trough within a trough" maximizes reagent recovery
- Reservoir has Pour-off spouts on all four corners
- Wide base provides stability and avoids spills
- 25 ml Reservoir has a divider accommodates eight channel pipets on one side and one to four channel pipets on the other
- Offered in bulk cases or individually wrapped (ideal for use in clean rooms)
The Sidecar Reservoir features “Two-in-One design” for precise reagent adding/mixing and “Sidecar” holds as little as 10 ml of liquid or as much as 20 ml. It has a ideal and efficient to use with small volume control and large volume test reagents.