Spécifications
- Description:Well Plate Stand
- Cat. no.:102096-948
- Supplier No.:70490-20
Spécifications
A propos de cet article
The well orienter is used for easy pipetting and identification of samples in microplates. With the well plate stand samples are viewed and pipetted at a comfortable 30° without strain.
- Convenient visual aid for pipetting and identification of samples in microplates
- Sharp, alpha-numeric characters compatible with 96-, 48- or 24-well microplates
- Neoprene pad and rubber feet prevents slippage of the plate and stand
- No more cross-referencing headaches
The well pad is a convenient note pad that corresponds to the layout of a 96-well plate. It is used to keep accurate records of samples in a microplate.