ContactUsLinkComponent
96 Well Orienter 2/Pk
Well Orienter, Pad and Stand, Electron Microscopy Sciences
Catalog # 102096-946
96 Well Orienter 2/Pk
Well Orienter, Pad and Stand, Electron Microscopy Sciences
Catalog # 102096-946
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Multi-colored 96 Well Orienter
  • Cat. no.:
    102096-946
  • Supplier No.:
    70490-18
Specifications
About this item
The well orienter is used for easy pipetting and identification of samples in microplates. With the well plate stand samples are viewed and pipetted at a comfortable 30° without strain.
  • Convenient visual aid for pipetting and identification of samples in microplates
  • Sharp, alpha-numeric characters compatible with 96-, 48- or 24-well microplates
  • Neoprene pad and rubber feet prevents slippage of the plate and stand
  • No more cross-referencing headaches
The well pad is a convenient note pad that corresponds to the layout of a 96-well plate. It is used to keep accurate records of samples in a microplate.
Customers Who Bought This Also Bought