ContactUsLinkComponent
Culture Cluster 6 Well 50/Pk
Costar® Brand Cell Culture Clusters, Electron Microscopy Sciences
# de catalogue 102095-520
Culture Cluster 6 Well 50/Pk
Costar® Brand Cell Culture Clusters, Electron Microscopy Sciences
# de catalogue 102095-520
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Costar Brand Cell Culture Clusters 6 Well
  • Cat. no.:
    102095-520
  • Supplier No.:
    64705-01
Spécifications
A propos de cet article
Tissue culture plates are designed and treated for a wide range of applications. Plates are made from optically clear, medical grade polystyrene with flat bottoms of uniform thickness, allowing for distortion-free observation and microscopic examination. Alphanumeric coordinates are placed on the same focal plane as cell growth-for easy referencing of well position. These plates are also for use with our NetWells and our MeshWell Plates.
  • Maintenance of stock cultures
  • Cell cloning
  • Hybridoma fusions
  • Scale up culture
  • Clone expansion
  • 3H-Thymidine
  • 51Cr Release assay
  • MTT assays
  • In vitro toxicology
  • Drug binding studies
  • Virus isolation
  • Viral plaque assay
  • Transformation
  • Carcinogenicity testing
  • Mutagenicity testing
Customers Who Bought This Also Bought