Spécifications
- Description:Costar Brand Cell Culture Clusters 6 Well
- Cat. no.:102095-520
- Supplier No.:64705-01
Spécifications
A propos de cet article
Tissue culture plates are designed and treated for a wide range of applications. Plates are made from optically clear, medical grade polystyrene with flat bottoms of uniform thickness, allowing for distortion-free observation and microscopic examination. Alphanumeric coordinates are placed on the same focal plane as cell growth-for easy referencing of well position. These plates are also for use with our NetWells and our MeshWell Plates.
- Maintenance of stock cultures
- Cell cloning
- Hybridoma fusions
- Scale up culture
- Clone expansion
- 3H-Thymidine
- 51Cr Release assay
- MTT assays
- In vitro toxicology
- Drug binding studies
- Virus isolation
- Viral plaque assay
- Transformation
- Carcinogenicity testing
- Mutagenicity testing