Spécifications
- Color:Green
- Description:380 Adhesion Slides
- Cat. no.:102094-800
- Supplier No.:63700-G1
Spécifications
A propos de cet article
TruBond™ adhesive slides have proven performance under high temperature and pH conditions. These adhesive slides work well with automatic IHC applications. The quality of these slides will exceed your expectations.