ContactUsLinkComponent
Trbnd Adhs Slde 380 100/Bx
TruBond™ Adhesion Slides, Electron Microscopy Sciences
# de catalogue 102094-800
Trbnd Adhs Slde 380 100/Bx
TruBond™ Adhesion Slides, Electron Microscopy Sciences
# de catalogue 102094-800
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    Green
  • Description:
    380 Adhesion Slides
  • Cat. no.:
    102094-800
  • Supplier No.:
    63700-G1
Spécifications
A propos de cet article
TruBond™ adhesive slides have proven performance under high temperature and pH conditions. These adhesive slides work well with automatic IHC applications. The quality of these slides will exceed your expectations.
Ordering information: All styles available in white, blue, green, and yellow. Pink is available in TruBond™ 360 and 200 only.
Packaging: TruBond™ is packaged 100 slides per box, 10 boxes to a case.
Customers Who Bought This Also Bought