To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Color:Clear
- Description:Maxisorp™ 96 Well Plates, PS
- Cat. no.:102094-770
- Application:Protein & antibody multiplexing, chemiluminescence
- Supplier No.:63494-CL
Spécifications
A propos de cet article
NUNC MaxiSorp plates have been the certified* ELISA plate industry standard for years. This same surface now sets a new standard for protein multiplexing.
- Hydrophilic MaxiSorp™ surface binds antibodies, protein and biomolecules
- Flat bottom, solid polystyrene plates
- Excellent for chemiluminescent, fluorescent and luminescent arrays
- Standard plate footprint is compatible with exiting readers
* Certified for absorption of IgG; CV of less than 5% between wells. All results are ±10% from the mean and background of all well ±0.005 absorbance units from the mean.